accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

AlH3COOH3COOH3COO

CAS RN 637-12-7

IUPAC Name: aluminium(3+) trioctadecanoate
Synonyms: aluminum stearate octadecanoic acid, aluminum salt aluminium tristearate aluminum tristearate aluminum(III) stearate
Molecular Weight (g/mol): 877.41
Molecular Formula: C54H105AlO6
InChi Key: CEGOLXSVJUTHNZ-UHFFFAOYSA-K
SMILES: [Al+3].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results