accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 637-60-5

IUPAC Name: hydrogen (4-methylphenyl)hydrazine chloride
Synonyms: hydrogen 4-methylphenylhydrazine chloride
Molecular Weight (g/mol): 158.63
Molecular Formula: C7H11ClN2
InChi Key: HMHWNJGOHUYVMD-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=C(NN)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results