accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 637-89-8

IUPAC Name: 4-sulfanylphenol
Synonyms: 4-sulfanylphenol
Molecular Weight (g/mol): 126.17
Molecular Formula: C6H6OS
InChi Key: BXAVKNRWVKUTLY-UHFFFAOYSA-N
SMILES: OC1=CC=C(S)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)