accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 63744-22-9

IUPAC Name: 6,8-dibromoimidazo[1,2-a]pyrazine
Synonyms: 6,8-dibromoimidazo[1,2-a]pyrazine
Molecular Weight (g/mol): 276.92
Molecular Formula: C6H3Br2N3
InChi Key: UQCZZGIPIMJBCL-UHFFFAOYSA-N
SMILES: BrC1=CN2C=CN=C2C(Br)=N1
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)