accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOSOOCl

CAS RN 63758-12-3

IUPAC Name: 2,3-dihydro-1,4-benzodioxine-6-sulfonyl chloride
Synonyms: 2,3-dihydro-1,4-benzodioxine-6-sulfonyl chloride
Molecular Weight (g/mol): 234.65
Molecular Formula: C8H7ClO4S
InChi Key: JWHSRWUHRLYAPM-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC2=C(OCCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)