accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3FHO

CAS RN 63762-79-8

IUPAC Name: 2-fluoro-5-methylphenol
Synonyms: 2-fluoro-5-methylphenol
Molecular Weight (g/mol): 126.13
Molecular Formula: C7H7FO
InChi Key: XEHPMVZYZDQLDN-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C(O)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results