accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 638-02-8

IUPAC Name: 2,5-dimethylthiophene
Synonyms: 2,5-dimethylthiophene
Molecular Weight (g/mol): 112.19
Molecular Formula: C6H8S
InChi Key: GWQOOADXMVQEFT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)S1
Boiling Point 
  • (5)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)