accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NOOHO

CAS RN 638-32-4

IUPAC Name: 3-carbamoylpropanoic acid
Synonyms: 3-carbamoylpropanoic acid
Molecular Weight (g/mol): 117.1
Molecular Formula: C4H7NO3
InChi Key: JDVPQXZIJDEHAN-UHFFFAOYSA-N
SMILES: NC(=O)CCC(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results