accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 638-38-0

IUPAC Name: manganese(2+) diacetate
Synonyms: manganese(II) acetate
Molecular Weight (g/mol): 173.03
Molecular Formula: C4H6MnO4
InChi Key: UOGMEBQRZBEZQT-UHFFFAOYSA-L
SMILES: [Mn++].CC([O-])=O.CC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results