accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 63812-15-7

CH3CH3OOHF

CAS RN 63812-15-7

IUPAC Name: 2-fluoro-2-methylpropanoic acid
Synonyms: 2-fluoro-2-methylpropanoic acid
Molecular Weight (g/mol): 106.1
Molecular Formula: C4H7FO2
InChi Key: NZDOWZQRNZLBOY-UHFFFAOYSA-N
SMILES: CC(C)(F)C(O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results