accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 639858-45-0

NH2FFOOH

CAS RN 639858-45-0

IUPAC Name: 5-amino-2,4-difluorobenzoic acid
Synonyms: 5-amino-2,4-difluorobenzoic acid
Molecular Weight (g/mol): 173.12
Molecular Formula: C7H5F2NO2
InChi Key: AMOPVOVLPZDNMD-UHFFFAOYSA-N
SMILES: NC1=C(F)C=C(F)C(=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results