accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

PSOOONa

CAS RN 63995-75-5

IUPAC Name: sodium 3-(diphenylphosphanyl)benzene-1-sulfonate
Synonyms: sodium 3-(diphenylphosphanyl)benzenesulfonate
Molecular Weight (g/mol): 364.33
Molecular Formula: C18H14NaO3PS
InChi Key: NTUROZDXWLPVHB-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=CC(=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results