accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 64-10-8

IUPAC Name: phenylurea
Synonyms: phenylurea
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: LUBJCRLGQSPQNN-UHFFFAOYSA-N
SMILES: NC(=O)NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results