CAS RN 64-17-5
Ethanol 99%+, Absolute, Extra Pure, SLR, Fisher Chemical™
CAS: 64-17-5 Molecular Formula: C2H6O MDL Number: 3568 InChI Key: LFQSCWFLJHTTHZ-UHFFFAOYSA-N Synonym: ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol PubChem CID: 702 ChEBI: CHEBI:16236 IUPAC Name: ethanol SMILES: CCO
Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™
CAS: 64-17-5 Molecular Formula: C2H6O InChI Key: LFQSCWFLJHTTHZ-UHFFFAOYSA-N Synonym: ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol PubChem CID: 702 ChEBI: CHEBI:16236 IUPAC Name: ethanol SMILES: CCO
Ethanol, 96%, technical, not denaturated
Ethanol, 99.5%, Extra Dry, absolute, AcroSeal™
CAS: 64-17-5 Molecular Formula: C2H6O Molecular Weight (g/mol): 46.069 MDL Number: MFCD00003568 InChI Key: LFQSCWFLJHTTHZ-UHFFFAOYSA-N Synonym: ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol PubChem CID: 702 ChEBI: CHEBI:16236 IUPAC Name: ethanol SMILES: CCO
Ethanol, 99.8%, technical, not denaturated
Ethanol, 96%, extra pure, denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex™
CAS: 64-17-5 Molecular Formula: C2H6O Molecular Weight (g/mol): 46.07 InChI Key: LFQSCWFLJHTTHZ-UHFFFAOYSA-N IUPAC Name: ethanol SMILES: CCO
Ethanol, 99.8%, anhyd.,denat. with 1%iPr-OH,1%MEK,0.001% Bitrex™
CAS: 64-17-5 Molecular Formula: C2H6O Molecular Weight (g/mol): 46.07 InChI Key: LFQSCWFLJHTTHZ-UHFFFAOYSA-N IUPAC Name: ethanol SMILES: CCO
Ethanol Absolute, for HPLC, Fisher Chemical™
CAS: 64-17-5 Molecular Formula: C2H6O MDL Number: 3568 InChI Key: LFQSCWFLJHTTHZ-UHFFFAOYSA-N PubChem CID: 702 ChEBI: CHEBI:16236 IUPAC Name: ethanol SMILES: CCO
Ethanol Absolute 99.8+%, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
CAS: 64-17-5 Molecular Formula: C2H6O Molecular Weight (g/mol): 46.069 MDL Number: 3568 InChI Key: LFQSCWFLJHTTHZ-UHFFFAOYSA-N PubChem CID: 702 ChEBI: CHEBI:16236 IUPAC Name: ethanol SMILES: CCO
Ethanol, 200 Proof (100%), USP/EP/ACS, Tax-Paid, Fisher Chemical™
CAS: 64-17-5 Molecular Formula: C2H6O Molecular Weight (g/mol): 46.07 InChI Key: LFQSCWFLJHTTHZ-UHFFFAOYSA-N Synonym: ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol PubChem CID: 702 ChEBI: CHEBI:16236 IUPAC Name: ethanol SMILES: CCO
Ethanol denatured (Untaxed), UK IDA standard, for license holders in Ireland and United Kingdom only, Solstice
CAS: 64-17-5 Molecular Formula: C2H6O Molecular Weight (g/mol): 46.069 InChI Key: LFQSCWFLJHTTHZ-UHFFFAOYSA-N Synonym: ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol PubChem CID: 702 ChEBI: CHEBI:16236 IUPAC Name: ethanol SMILES: CCO
HYDRANAL™-Buffer Base, Solstice
Liquid buffer medium for Karl Fischer titration in alkaline samples (based on salicylic acid)
HYDRANAL™-Solvent E, Solstice
Medium for volumetric two-component Karl Fischer titration (ethanol-based)