accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrCH3NCH3CH3H3C

CAS RN 64-20-0

IUPAC Name: tetramethylazanium bromide
Synonyms: tetramethylammonium bromide TMAB
Molecular Weight (g/mol): 154.05
Molecular Formula: C4H12BrN
InChi Key: DDFYFBUWEBINLX-UHFFFAOYSA-M
SMILES: [Br-].C[N+](C)(C)C
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)