accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OSOOOH3C

CAS RN 64-67-5

IUPAC Name: diethyl sulfate
Synonyms: diethyl sulfate diaethylsulfat diethylsulfate ccris 242 diethylester kyseliny sirove diethyl tetraoxosulfate
Molecular Weight (g/mol): 154.18
Molecular Formula: C4H10O4S
InChi Key: DENRZWYUOJLTMF-UHFFFAOYSA-N
SMILES: CCOS(=O)(=O)OCC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)