CAS RN 640-19-7
IUPAC Name:
2-fluoroacetamide
Synonyms:
fluoroacetamide
Molecular Weight (g/mol):
77.06
Molecular Formula:
C2H4FNO
InChi Key:
FVTWJXMFYOXOKK-UHFFFAOYSA-N
SMILES:
NC(=O)CF
1
–
1
of
1
results
1
Fluoroacetamide, TRC
CAS: 640-19-7 Molecular Formula: C2 H4 F N O Molecular Weight (g/mol): 77.06 Synonym: FAA,Acetamide, fluoro- IUPAC Name: 2-fluoroacetamide SMILES: NC(=O)CF