accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONHSHN

CAS RN 64038-64-8

IUPAC Name: ethyl 2-sulfanylidene-2,3-dihydro-1H-imidazole-4-carboxylate
Synonyms: ethyl 2-sulfanylidene-1,3-dihydroimidazole-4-carboxylate
Molecular Weight (g/mol): 172.2
Molecular Formula: C6H8N2O2S
InChi Key: PONOGPICUOALRU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CNC(=S)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results