accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SnH3CCH3H3C

CAS RN 64099-82-7

IUPAC Name: tributyl(prop-1-yn-1-yl)stannane
Synonyms: tributyl(prop-1-yn-1-yl)stannane
Molecular Weight (g/mol): 329.12
Molecular Formula: C15H30Sn
InChi Key: KCQJLTOSSVXOCC-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C#CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results