accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 641-70-3

IUPAC Name: 4-nitro-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 4-nitro-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 193.11
Molecular Formula: C8H3NO5
InChi Key: ROFZMKDROVBLNY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results