CAS RN 64169-58-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Chlorocitalopram, Hydrobromide, TRC
CAS: 64169-58-0 Molecular Formula: C20 H21 Cl N2 O . Br H Molecular Weight (g/mol): 421.76 IUPAC Name: 1-(4-chlorophenyl)-1-[3-(dimethylamino)propyl]-3H-2-benzofuran-5-carbonitrile;hydrobromide SMILES: Br.CN(C)CCCC1(OCc2cc(ccc12)C#N)c3ccc(Cl)cc3