accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6418-38-8

IUPAC Name: 2,3-difluorophenol
Synonyms: 2,3-difluorophenol
Molecular Weight (g/mol): 130.09
Molecular Formula: C6H4F2O
InChi Key: RPEPGIOVXBBUMJ-UHFFFAOYSA-N
SMILES: OC1=CC=CC(F)=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results