accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 64183-27-3

IUPAC Name: 5-(6-amino-9H-purin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
Synonyms: 5-(6-aminopurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
Molecular Weight (g/mol): 269.24
Molecular Formula: C10H12FN5O3
InChi Key: ZGYYPTJWJBEXBC-UHFFFAOYNA-N
SMILES: NC1=C2N=CN(C3OC(CO)C(O)C3F)C2=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)