accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3

CAS RN 643-58-3

IUPAC Name: 2-methyl-1,1'-biphenyl
Synonyms: 2-methylbiphenyl
Molecular Weight (g/mol): 168.24
Molecular Formula: C13H12
InChi Key: ALLIZEAXNXSFGD-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results