CAS RN 64316-11-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Chlorodibromoacetaldehyde (>90%), TRC
CAS: 64316-11-6 Molecular Formula: C2 H Br2 Cl O Molecular Weight (g/mol): 236.29 Synonym: 2,2-Dibromo-2-chloro-acetaldehyde,Chlorodibromoacetaldehyde,Dibromochloroacetaldehyde,Dibromochloro-acetaldehyde IUPAC Name: 2,2-dibromo-2-chloroacetaldehyde SMILES: ClC(Br)(Br)C=O