accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OO(R)(R)OOOOO

CAS RN 64339-95-3

IUPAC Name: 4-(benzoyloxy)-2,5-dioxooxolan-3-yl benzoate
Synonyms: 4-(benzoyloxy)-2,5-dioxooxolan-3-yl benzoate
Molecular Weight (g/mol): 340.29
Molecular Formula: C18H12O7
InChi Key: OXIKRMSPXYQFOT-UHFFFAOYNA-N
SMILES: O=C(OC1C(OC(=O)C2=CC=CC=C2)C(=O)OC1=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results