accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 644-06-4

IUPAC Name: 6,7-dimethoxy-2,2-dimethyl-2H-chromene
Synonyms: precocene II precocene 2 precocen 2 ageratochromene pricocene ii pricocene II
Molecular Weight (g/mol): 220.27
Molecular Formula: C13H16O3
InChi Key: PTIDGSWTMLSGAH-UHFFFAOYSA-N
SMILES: COC1=CC2=C(C=CC(C)(C)O2)C=C1OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results