accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 644-08-6

CH3

CAS RN 644-08-6

IUPAC Name: 4-methyl-1,1'-biphenyl
Synonyms: 4-methyl-1,1'-biphenyl
Molecular Weight (g/mol): 168.24
Molecular Formula: C13H12
InChi Key: ZZLCFHIKESPLTH-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results