accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHHOHONHNHHOHOHO

CAS RN 64431-96-5

IUPAC Name: 2-[(3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}propyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Synonyms: bis-tris propane
Molecular Weight (g/mol): 282.34
Molecular Formula: C11H26N2O6
InChi Key: HHKZCCWKTZRCCL-UHFFFAOYSA-N
SMILES: OCC(CO)(CO)NCCCNC(CO)(CO)CO
Quantity 
  • (1)
  • (3)
  • (6)
  • (2)
  • (1)
  • (6)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (15)
Physical Form 
  • (4)
  • (12)
  • (3)
pH 
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (15)
  • (2)
  • (1)
  • (3)
  • (6)
  • (2)
  • (1)
  • (6)
  • (2)
  • (4)
  • (15)
  • (4)
  • (12)
  • (3)
  • (12)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (4)