accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 645-12-5

IUPAC Name: 5-nitrofuran-2-carboxylic acid
Synonyms: 5-nitro-2-furoic acid
Molecular Weight (g/mol): 157.08
Molecular Formula: C5H3NO5
InChi Key: IODMEDPPCXSFLD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(O1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)