accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 645-54-5

IUPAC Name: 2-phenylethanethioamide
Synonyms: 2-phenylethanethioamide
Molecular Weight (g/mol): 151.23
Molecular Formula: C8H9NS
InChi Key: CJXBHFANXQMZBF-UHFFFAOYSA-N
SMILES: NC(=S)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results