accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NONHOHN

CAS RN 645-93-2

IUPAC Name: 6-amino-1,2,3,4-tetrahydro-1,3,5-triazine-2,4-dione
Synonyms: ammelide
Molecular Weight (g/mol): 128.09
Molecular Formula: C3H4N4O2
InChi Key: YSKUZVBSHIWEFK-UHFFFAOYSA-N
SMILES: NC1=NC(=O)NC(=O)N1
Quantity 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results