accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHO

CAS RN 646-30-0

IUPAC Name: nonadecanoic acid
Synonyms: nonadecanoic acid
Molecular Weight (g/mol): 298.51
Molecular Formula: C19H38O2
InChi Key: ISYWECDDZWTKFF-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)