accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 647-42-7

IUPAC Name: 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctan-1-ol
Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctan-1-ol
Molecular Weight (g/mol): 364.11
Molecular Formula: C8H5F13O
InChi Key: GRJRKPMIRMSBNK-UHFFFAOYSA-N
SMILES: OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results