CAS RN 64742-49-0
Petroleum Ether 40-60°C, Extra Pure, SLR, Fisher Chemical™
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum Ether 40-60°C, Certified AR for Analysis, N-Hexane <2%, Fisher Chemical™
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum Ether 60-80°C, Extra Pure, SLR, Fisher Chemical™
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum Ether 100-120°C, Extra Pure, SLR, Fisher Chemical™
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum Ether 100-120°C, Certified AR for Analysis, N-Hexane <2%, Fisher Chemical™
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum ether, ACS reagent, boiling range 40-60°C
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum ether, for analysis, boiling range 80-110°C
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum ether, extra pure, boiling range 40-60°C, Thermo Scientific Chemicals
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum Ether 60-80°C, Certified AR for Analysis, N-Hexane <50%, Fisher Chemical™
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 IUPAC Name: Petroleum ether SMILES: CCCC(C)C
Petroleum ether, for residue analysis, boiling range 40-60°C, ECD tested halocarbons free
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 SMILES: CCCC(C)C
Petroleum Ether 40-60C CertiFied for analysis , Fisher Chemical™
Petroleum ether 40-60C CertiFied for analysis 200L n-hexane <2%
Petroleum Ether, Optima™ for HPLC and GC, Fisher Chemical™
Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N PubChem CID: 7892 IUPAC Name: 2-methylpentane SMILES: CCCC(C)C
Petroleum ether, for residue analysis, 40-60°C, for trace anal. of polyarom. hydrocarbons
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 IUPAC Name: Petroleum ether SMILES: CCCC(C)C
Petroleum ether, ACS reagent, boiling range 35-60°C
CAS: 64742-49-0 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.178 MDL Number: MFCD00081849 InChI Key: AFABGHUZZDYHJO-UHFFFAOYSA-N Synonym: isohexane,pentane, 2-methyl,2-methylpentan,2-methyl pentane,dimethylpropylmethane,1,1-dimethylbutane,iso-hexane,methyl pentane,unii-49ib0u6mld,2-methyl-pentane PubChem CID: 7892 IUPAC Name: Petroleum ether SMILES: CCCC(C)C
Heptane, Extra Pure, Fraction, SLR, Fisher Chemical™
CAS: 64742-49-0 Molecular Formula: C7H16 Molecular Weight (g/mol): 100.21 MDL Number: MFCD00081849 InChI Key: IMNFDUFMRHMDMM-UHFFFAOYSA-N PubChem CID: 8900 ChEBI: CHEBI:43098 SMILES: CCCCCCC
- 1
- 2