accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 64744-50-9

IUPAC Name: 2-azaspiro[4.5]decan-3-one
Synonyms: 2-azaspiro[4.5]decan-3-one
Molecular Weight (g/mol): 153.23
Molecular Formula: C9H15NO
InChi Key: JAWPQJDOQPSNIQ-UHFFFAOYSA-N
SMILES: O=C1CC2(CN1)CCCCC2
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results