accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHNHCH3

CAS RN 6483-50-7

IUPAC Name: 1,4-diethylpiperazine-1,4-diium
Synonyms: 1,4-diethylpiperazine-1,4-diium
Molecular Weight (g/mol): 144.26
Molecular Formula: C8H20N2
InChi Key: DDPRYTUJYNYJKV-UHFFFAOYSA-P
SMILES: CC[NH+]1CC[NH+](CC)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results