accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 64872-77-1

IUPAC Name: 1-[4-(4-chlorophenyl)-2-[(2,6-dichlorophenyl)sulfanyl]butyl]-1H-imidazole; nitric acid
Synonyms: nitrato de butoconazol
Molecular Weight (g/mol): 474.78
Molecular Formula: C19H18Cl3N3O3S
InChi Key: ZHPWRQIPPNZNML-UHFFFAOYNA-N
SMILES: O[N+]([O-])=O.ClC1=CC=C(CCC(CN2C=CN=C2)SC2=C(Cl)C=CC=C2Cl)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Certifications/Compliance 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)