accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 64896-28-2

H3C(S)(S)CH3PPPhPhPhPh

CAS RN 64896-28-2

IUPAC Name: [3-(diphenylphosphanyl)butan-2-yl]diphenylphosphane
Synonyms: chiraphos
Molecular Weight (g/mol): 426.48
Molecular Formula: C28H28P2
InChi Key: FWXAUDSWDBGCMN-UHFFFAOYNA-N
SMILES: CC(C(C)P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results