accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6493-05-6

H3CNNNCH3ONCH3OO

CAS RN 6493-05-6

IUPAC Name: 3,7-dimethyl-1-(5-oxohexyl)-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: pentoxifylline trental pentoxyfylline pentoxiphylline pentoxil oxpentifylline pentoxifilina pentoxyphylline pentoxifyllin torental
Molecular Weight (g/mol): 278.31
Molecular Formula: C13H18N4O3
InChi Key: BYPFEZZEUUWMEJ-UHFFFAOYSA-N
SMILES: CN1C=NC2=C1C(=O)N(CCCCC(C)=O)C(=O)N2C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results