accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOOH3C

CAS RN 6496-89-5

IUPAC Name: 2-(2,4-dimethoxyphenyl)acetate
Synonyms: 2-(2,4-dimethoxyphenyl)acetate
Molecular Weight (g/mol): 195.2
Molecular Formula: C10H11O4
InChi Key: ZFXFMGARFHRTTO-UHFFFAOYSA-M
SMILES: COC1=CC=C(CC([O-])=O)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results