accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOSNNH2

CAS RN 64987-08-2

IUPAC Name: ethyl 2-(2-amino-1,3-thiazol-4-yl)-2-oxoacetate
Synonyms: ethyl 2-(2-amino-1,3-thiazol-4-yl)-2-oxoacetate
Molecular Weight (g/mol): 200.21
Molecular Formula: C7H8N2O3S
InChi Key: XNVRKLCQBZTGNA-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)C1=CSC(N)=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)