CAS RN 65-22-5
Pyridoxal hydrochloride, 99%
CAS: 65-22-5 Molecular Formula: C8H10ClNO3 Molecular Weight (g/mol): 203.622 MDL Number: MFCD00012809 InChI Key: FCHXJFJNDJXENQ-UHFFFAOYSA-N Synonym: pyridoxal hydrochloride,pyridoxal hcl,vitamin b6 hydrochloride,3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride,unii-1416kf0qbc,2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride,pl hcl,3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride,3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride,3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride PubChem CID: 6171 IUPAC Name: 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride SMILES: CC1=NC=C(C(=C1O)C=O)CO.Cl
Pyridoxal hydrochloride, 99%
CAS: 65-22-5 Molecular Formula: C8H9NO3·HCl Molecular Weight (g/mol): 203.62 MDL Number: MFCD00012809 InChI Key: FCHXJFJNDJXENQ-UHFFFAOYSA-N Synonym: pyridoxal hydrochloride,pyridoxal hcl,vitamin b6 hydrochloride,3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride,unii-1416kf0qbc,2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride,pl hcl,3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride,3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride,3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride PubChem CID: 6171 IUPAC Name: 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride SMILES: CC1=NC=C(C(=C1O)C=O)CO.Cl
Melford Pyridoxal HCl
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Pyridoxal Hydrochloride, TRC
CAS: 65-22-5 Molecular Formula: C8 H9 N O3 . Cl H Molecular Weight (g/mol): 203.62 Synonym: 4-Pyridinecarboxaldehyde, 3-hydroxy-5-(hydroxymethyl)-2-methyl-, hydrochloride (9CI),Pyridoxal, hydrochloride (7CI,8CI),2-Methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride,3-Hydroxy-5-(hydroxymethyl)-2-methyl-4-pyridinecarboxaldehyde hydrochloride,3-Hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride IUPAC Name: 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride SMILES: Cl.Cc1ncc(CO)c(C=O)c1O
Pyridoxal Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Pyridoxal Hydrochloride, Fisher BioReagents
CAS: 65-22-5 Molecular Formula: C8H10ClNO3 Molecular Weight (g/mol): 203.622 InChI Key: FCHXJFJNDJXENQ-UHFFFAOYSA-N Synonym: pyridoxal hydrochloride,pyridoxal hcl,vitamin b6 hydrochloride,3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride,unii-1416kf0qbc,2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride,pl hcl,3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride,3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride,3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride PubChem CID: 6171 IUPAC Name: 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride SMILES: CC1=NC=C(C(=C1O)C=O)CO.Cl
Pyridoxal hydrochloride 100 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality