accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 65-82-7

H3CS(S)NHCH3OOHO

CAS RN 65-82-7

IUPAC Name: 2-acetamido-4-(methylsulfanyl)butanoic acid
Synonyms: L-methionine, N-acetyl-
Molecular Weight (g/mol): 191.25
Molecular Formula: C7H13NO3S
InChi Key: XUYPXLNMDZIRQH-UHFFFAOYNA-N
SMILES: CSCCC(NC(C)=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)