accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 65-86-1

IUPAC Name: 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid
Synonyms: orotic acid 奥拉米特 orotsaeure orazamide
Molecular Weight (g/mol): 156.1
Molecular Formula: C5H4N2O4
InChi Key: PXQPEWDEAKTCGB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=O)NC(=O)N1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results