accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 65001-21-0

NBrSOOCl

CAS RN 65001-21-0

IUPAC Name: 5-bromopyridine-3-sulfonyl chloride
Synonyms: 5-bromopyridine-3-sulfonyl chloride
Molecular Weight (g/mol): 256.5
Molecular Formula: C5H3BrClNO2S
InChi Key: AVILRJQPDYPXFQ-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC(Br)=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results