accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NHNF

CAS RN 65020-70-4

IUPAC Name: 5-(4-fluorophenyl)-1H-imidazole
Synonyms: 4-(4-fluorophenyl)-3H-imidazole
Molecular Weight (g/mol): 162.17
Molecular Formula: C9H7FN2
InChi Key: QYDAFWKRBZYBOB-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1=CN=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results