accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 65039-08-9

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium bromide
Synonyms: 1-ethyl-3-methylimidazolium bromide
Molecular Weight (g/mol): 191.07
Molecular Formula: C6H11BrN2
InChi Key: GWQYPLXGJIXMMV-UHFFFAOYSA-M
SMILES: [Br-].CCN1C=C[N+](C)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)