accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COPOOCH3FFBr

CAS RN 65094-22-6

IUPAC Name: diethyl (bromodifluoromethyl)phosphonate
Synonyms: diethyl bromodifluoromethylphosphonate
Molecular Weight (g/mol): 267.01
Molecular Formula: C5H10BrF2O3P
InChi Key: QRADKVYIJIAENZ-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)C(F)(F)Br
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (3)
  • (7)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)