accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOFFOHFF

CAS RN 652-34-6

IUPAC Name: 2,3,5,6-tetrafluoro-4-hydroxybenzoic acid
Synonyms: 2,3,5,6-tetrafluoro-4-hydroxybenzoic acid
Molecular Weight (g/mol): 210.08
Molecular Formula: C7H2F4O3
InChi Key: FTLHGQOBAPTEHE-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(F)=C(O)C(F)=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results